Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0183

Sigma-Aldrich

EM20-25

≥98% (HPLC)

Synonym(s):

5-(6-Chloro-2,4-dioxo-1,3,4,10-tetrahydro-2H-9-oxa-1,3-diaza-anthracen-10-yl)-pyrimidine-2,4,6-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9ClN4O6
CAS Number:
Molecular Weight:
376.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

light yellow to yellow

solubility

DMSO: ≥5 mg/mL at ~60 °C

storage temp.

2-8°C

SMILES string

Clc1ccc2OC3=C(C(C4C(=O)NC(=O)NC4=O)c2c1)C(=O)NC(=O)N3

InChI

1S/C15H9ClN4O6/c16-4-1-2-6-5(3-4)7(8-10(21)17-14(24)18-11(8)22)9-12(23)19-15(25)20-13(9)26-6/h1-3,7-8H,(H2,19,20,23,25)(H2,17,18,21,22,24)

InChI key

GGEVZGGAQHNWQN-UHFFFAOYSA-N

Application

EM20-25 was screened among other anti-cancer drugs to evaluate the effect on mitochondrial membrane potential in human prostate cancer cells and healthy mouse liver tissue.

Biochem/physiol Actions

EM20-25 disrupts the BCL-2/BAX interactions and activates caspase-9 in cells overexpressing BCL-2. It sensitizes the BCL-2 expressing leukemic cells to the effects of chemotherapy involving staurosporine and chlorambucil.
EM20-25 is an analog of HA14-1 that antagonizes the effects of the anti-apoptotic protein BCL-2, and causes opening of the mitochondrial permeability transition pore. Unlike HA14-1, EM20-25 does not effect mitochondrial respiration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nelly Buron et al.
PloS one, 5(3), e9924-e9924 (2010-04-03)
Current limitations of chemotherapy include toxicity on healthy tissues and multidrug resistance of malignant cells. A number of recent anti-cancer strategies aim at targeting the mitochondrial apoptotic machinery to induce tumor cell death. In this study, we set up protocols
Aayushi Singh et al.
Biomolecular concepts, 12(1), 94-109 (2021-07-26)
We previously reported that M. tb on its own as well as together with HIV inhibits macrophage apoptosis by upregulating the expression of Bcl2 and Inhibitor of Apoptosis (IAP). In addition, recent reports from our lab showed that stimulation of
Eva Milanesi et al.
The Journal of biological chemistry, 281(15), 10066-10072 (2006-02-17)
We have investigated the mitochondrial effects of BH3I-2', Chelerythrine, and HA14-1, small organic molecules that share the ability to bind the BH3 domain of BCL-2. All compounds displayed a biphasic effect on mitochondrial respiration with uncoupling at low concentrations and
Dennis Ma et al.
Scientific reports, 7, 42957-42957 (2017-02-22)
Enhanced mitochondrial stability and decreased dependence on oxidative phosphorylation confer an acquired resistance to apoptosis in cancer cells, but may present opportunities for therapeutic intervention. The compound pancratistatin (PST) has been shown to selectively induce apoptosis in cancer cells. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service