Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0050

Sigma-Aldrich

CGP 37849

≥98% (HPLC)

Synonym(s):

(3E)-2-Amino-4-methyl-5-phosphono-3-pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NO5P
CAS Number:
Molecular Weight:
209.14
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: ≥2 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

C\C(CP(O)(O)=O)=C/C(N)C(O)=O

InChI

1S/C6H12NO5P/c1-4(3-13(10,11)12)2-5(7)6(8)9/h2,5H,3,7H2,1H3,(H,8,9)(H2,10,11,12)/b4-2+

InChI key

BDYHNCZIGYIOGJ-DUXPYHPUSA-N

Application

CGP 37849 may be used in NMDA-mediated cell signaling studies.

Biochem/physiol Actions

CGP 37849 induces phencyclidine-like behavioral syndrome in amygdala-kindled rats that includes ataxia, locomotion and stereotypies.[1]
CGP 37849 is a potent orally active competitive NMDA antagonist and anticonvulsant.
Potent orally active competitive NMDA antagonist and anticonvulsant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krzysztof Wedzony et al.
Pharmacological reports : PR, 60(6), 856-864 (2009-02-13)
Postnatal hypofunction of N-methyl-D-aspartate (NMDA) receptors leads to several behavioral deficits in adult rats resembling deficits typical of schizophrenia-like deficits of sensorimotor gating. Thus far, it is not known whether the above disruptions are sensitive to neuroleptic drugs. In order
A Y Gemperle et al.
Neuroscience, 117(3), 681-695 (2003-03-06)
The mode of action of the antipsychotic drugs clozapine, haloperidol and iloperidone was investigated in layer V of prefrontal cortex slices using extracellular field potential, intracellular sharp-electrode as well as whole-cell voltage clamp recording techniques. Intracellular investigations on a broad
K Wedzony et al.
Neuroscience, 153(4), 1370-1379 (2008-04-25)
The malfunction of glutamatergic neurotransmission in the neonatal or postnatal periods may be a risk factor for the appearance of neuroanatomical, neurochemical or functional changes that are characteristic of schizophrenia. Thus, the present study was undertaken to investigate whether blockade
K Wedzony et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 56(2), 205-221 (2005-06-30)
Malfunction of glutamatergic neurotransmission in postnatal period is considered to be a risk factor for development of schizophrenia. Thus, the present study investigates the impact of NMDA receptor blockade in the postnatal period on the density of tyrosine hydroxylase immunoreactive
Mélanie Langlois et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(49), 16523-16535 (2010-12-15)
Mesial temporal lobe epilepsy (MTLE) is characterized by focal seizures, associated with hippocampal sclerosis, and often resistance to antiepileptic drugs. The parafascicular nucleus (PF) of the thalamus is involved in the generation of physiological oscillatory rhythms. It receives excitatory inputs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service