Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB01002

Sigma-Aldrich

Guineensine

≥90% (LC/MS-ELSD)

Synonym(s):

(2E,4E,12E)-13-(1,3-benzodioxol-5-yl)-N-isobutyl-trideca-2,4,12-trienamide, Pipyahyine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33NO3
CAS Number:
Molecular Weight:
383.52
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

383.52

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C24H33NO3/c1-20(2)18-25-24(26)14-12-10-8-6-4-3-5-7-9-11-13-21-15-16-22-23(17-21)28-19-27-22/h8,10-17,20H,3-7,9,18-19H2,1-2H3,(H,25,26)/b10-8+,13-11+,14-12+

InChI key

FPMPOFBEYSSYDQ-AUVZEZIHSA-N

General description

Guineensine, also known as Pipyahyine, is an alkaloid and a member of benzodioxoles is a natural product commonly available from Piper Sp.(P. nigrum and P. longum and P. retrofractum) plants. Existing research suggests that this plant-derived metabolite exerts various biological activities, including antimicrobial, anticancer, anti-inflammatory, antiviral and antioxidant properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

According to the existing research, Guineensine inhibits endocannabinoid uptake, leading to significant anti-inflammatory and analgesic effects, and also interacts with various receptors, including dopamine transporter DAT, 5HT2A, and sigma receptors, suggesting potential polypharmacological activity.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruben Bartholomäus et al.
ChemMedChem, 14(17), 1590-1596 (2019-07-20)
Guineensine ((2E,4E,12E)-13-(benzo[d][1,3]dioxol-5-yl)-N-isobutyltrideca-2,4,12-trienamide) is a plant-derived natural product that inhibits reuptake of the endocannabinoid anandamide with sub-micromolar potency. We have established a highly efficient total synthesis of guineensine, which provided the natural product in only five steps from commercially available 3-nonyn-1-ol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service