Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00914

Sigma-Aldrich

Isovanillic Acid 3-Sulfate Disodium Salt

≥96% (HPLC)

Synonym(s):

Isovanillic acid 3-sulfate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6Na2O7S
CAS Number:
Molecular Weight:
292.17
UNSPSC Code:
12352205
NACRES:
NA.25

Assay

≥96% (HPLC)

form

powder

color

, White to off-white

solubility

water: 40 mg/mL

storage temp.

2-8°C

SMILES string

COC(C=C1)=C(OS(=O)(O[Na])=O)C=C1C(O[Na])=O

General description

Isovanillic acid 3-O-sulfate (IVAS) is a gut microbial phenolic metabolite primarily derived from consumed cyanidin 3-O-glucoside, a major compound in berry fruits. IVAS is the most effective among the gut microbial phenolic metabolites tested to enhance glucose uptake & metabolism in human skeletal muscle myoblast LHCN-M2. IIVAS is shown to increase glucose transport via GLUT4- and PI3K-dependent mechanisms. IVAS also upregulates GLUT1, GLUT4, PI3K p85a protein level, and Akt phosphorylation, establishing a novel link between gut microbiota, host diet and skeletal muscle energy source utilization.Also another study shows that IVAS restores glycosaminoglycans (GAGs) level and reduce inflammatory markers in diabetic human aortic endothelial cells (HAECs).

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service