Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00563

Sigma-Aldrich

Kipukasin D

≥90% (LC/MS-UV)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O9
CAS Number:
Molecular Weight:
422.39
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C(C1=C(C=C(C=C1C)OC)OC)O[C@@H]2[C@H](O[C@H]([C@@H]2O)N(C(N3)=O)C=CC3=O)CO

InChI

1S/C19H22N2O9/c1-9-6-10(27-2)7-11(28-3)14(9)18(25)30-16-12(8-22)29-17(15(16)24)21-5-4-13(23)20-19(21)26/h4-7,12,15-17,22,24H,8H2,1-3H3,(H,20,23,26)/t12-,15-,16-,17-/m1/s1

InChI key

LQYPUZKOEZWGBX-BASLNEPJSA-N

General description

Natural product derived from fungal source.

Biochem/physiol Actions

Antifungal and antibacterial activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Jiao et al.
Journal of natural products, 70(8), 1308-1311 (2007-07-05)
Seven new aroyl uridine derivatives (kipukasins A-G; 1-7) were isolated from solid-substrate fermentation cultures of two different Hawaiian isolates of Aspergillus versicolor. The structures of compounds 1-7 were determined by analysis of NMR and MS data. The nucleoside portion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service