Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00428

Sigma-Aldrich

Resibufogenin

≥98% (HPLC)

Synonym(s):

14,15-β-epoxy-3-β-hydroxy-5-β-bufa-20,22-dienolide, Bufogenin, RBG, Recibufogenin, Respigon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@@]12[C@@]3(O[C@@H]3C[C@@H]2C4=COC(C=C4)=O)[C@]5([H])CC[C@]6([H])C[C@@H](O)CC[C@]6(C)[C@@]5([H])CC1

InChI

1S/C24H32O4/c1-22-9-7-16(25)11-15(22)4-5-18-17(22)8-10-23(2)19(12-20-24(18,23)28-20)14-3-6-21(26)27-13-14/h3,6,13,15-20,25H,4-5,7-12H2,1-2H3/t15-,16+,17+,18-,19-,20-,22+,23-,24-/m1/s1

InChI key

ATLJNLYIJOCWJE-CWMZOUAVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Resibufogenin is isolated from the skin venom of the Asiatic toad (Bufo gargarizans) known as Chan su in Traditional Chinese Medicine.

Biochem/physiol Actions

Resibufogenin is a specific sodium/potassium-ATPase protein (Na+/K+-ATPase) inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service