Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00404

Sigma-Aldrich

β-Lac-PEG3-Azide

≥90%

Synonym(s):

(8-Azido-3,6-dioxaoctyl) β-D-galactopyranosyl-(1→4)-D-glucopyranoside, (8-Azido-3,6-dioxaoctyl) β-D-lactoopyranoside, 2-{2-[2-azidoethoxy]ethoxy}ethyl β-D-lactoopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H33N3O13
Molecular Weight:
499.47
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥90%

form

solid

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)[C@@H](CO)O2)[C@@H](CO)O[C@@H](OCCOCCOCCN=[N+]=[N-])[C@@H]1O

InChI

1S/C18H33N3O13/c19-21-20-1-2-29-3-4-30-5-6-31-17-15(28)13(26)16(10(8-23)33-17)34-18-14(27)12(25)11(24)9(7-22)32-18/h9-18,22-28H,1-8H2/t9?,10?,11-,12-,13+,14?,15?,16+,17+,18-/m0/s1

InChI key

ODCSAHBYDSBXRV-UYLHYHGYSA-N

General description

β-Lac-PEG3-Azide is a PEGylated glycoside.

Biochem/physiol Actions

β-Lac-PEG3-Azide is a PEGylated glycoside. PEGylated glycosides are used as ligands for Click Chemistry. They may also be important as carbohydrate carriers in glycotherapeutic applications, drug carriers for site-specific delivery, drug enhancers for decreasing toxicity, and solubility and protein enhancers.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service