Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00322

Sigma-Aldrich

Crotonoside

≥95% (HPLC)

Synonym(s):

Crotonoside, 2,3-Dihydro-2-Oxo-Adenosine, 9-(β-D-Ribofuranosyl)isoguanine, Isoguanine Riboside, Isoguanosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O5
CAS Number:
Molecular Weight:
283.24
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
To inquire about this product SMB00322, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

≥95% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

NC1=NC(NC2=C1N=CN2[C@H]3[C@H](O)[C@H](O)[C@@H](CO)O3)=O

InChI

1S/C10H13N5O5/c11-7-4-8(14-10(19)13-7)15(2-12-4)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)/t3-,5-,6-,9-/m1/s1

InChI key

MIKUYHXYGGJMLM-UUOKFMHZSA-N

General description

Crotonoside, or isoguanosine, is a naturally occurring active isomer of guanosine that is found in the seeds of Croton tiglium.

Biochem/physiol Actions

Crotonoside, or isoguanosine, is a naturally occurring active isomer of guanosine that is found in the seeds of Croton tiglium, which is used in Traditional Chinese Medicine. It has shown more activity than adenosine in reducing the blood pressure of rabbits and cats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service