Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00238

Sigma-Aldrich

Jarsuroside C

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H64O24
CAS Number:
Molecular Weight:
976.96
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)[C@@H]1CC(=O)OC[C@@H]3[C@H](C)[C@H](C[C@H]3C(CO)CO)OC(=O)C[C@H]4C(=C/C)\[C@@H](OC=C4C(=O)OC)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O

InChI

1S/C44H64O24/c1-6-20-23(26(39(57)59-4)16-62-41(20)67-43-37(55)35(53)33(51)29(13-47)65-43)9-31(49)61-15-25-18(3)28(8-22(25)19(11-45)12-46)64-32(50)10-24-21(7-2)42(63-17-27(24)40(58)60-5)68-44-38(56)36(54)34(52)30(14-48)66-44/h6-7,16-19,22-25,28-30,33-38,41-48,51-56H,8-15H2,1-5H3/b20-6+,21-7+/t18-,22-,23-,24-,25+,28-,29+,30+,33+,34+,35-,36-,37+,38+,41-,42-,43-,44-/m0/s1

InChI key

QGVKUNWDVOEJOC-DDFLQQJCSA-N

General description

Natural product derived from plant source.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service