Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00158

Sigma-Aldrich

Olivil 4′-O-β-D-glucopyranoside

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34O12
CAS Number:
Molecular Weight:
538.54
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc(C[C@@]2(O)CO[C@@H]([C@H]2CO)c3ccc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c(OC)c3)ccc1O

InChI

1S/C26H34O12/c1-34-18-7-13(3-5-16(18)29)9-26(33)12-36-24(15(26)10-27)14-4-6-17(19(8-14)35-2)37-25-23(32)22(31)21(30)20(11-28)38-25/h3-8,15,20-25,27-33H,9-12H2,1-2H3/t15-,20-,21-,22+,23-,24-,25-,26-/m1/s1

InChI key

OXHVZEZYYQQCRJ-CTQTXEDXSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service