Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00120

Sigma-Aldrich

Acoric acid

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O4
CAS Number:
Molecular Weight:
268.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC(C)C(=O)[C@]1(CC[C@@H](C)C(=O)C1)[C@H](C)CC(O)=O

InChI

1S/C15H24O4/c1-9(2)14(19)15(11(4)7-13(17)18)6-5-10(3)12(16)8-15/h9-11H,5-8H2,1-4H3,(H,17,18)/t10-,11-,15+/m1/s1

InChI key

ZIOCYJNRYIRTQD-HFAKWTLXSA-N

General description

Natural product derived from plant source.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service