Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00117

Sigma-Aldrich

Vernodalol

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O8
CAS Number:
Molecular Weight:
392.40
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COC(=O)C(=C)[C@H]1[C@H](O)[C@H]2C(=C)C(=O)OC[C@]2(C[C@@H]1OC(=O)C(=C)CO)C=C

InChI

1S/C20H24O8/c1-6-20-7-13(28-17(23)10(2)8-21)14(11(3)18(24)26-5)16(22)15(20)12(4)19(25)27-9-20/h6,13-16,21-22H,1-4,7-9H2,5H3/t13-,14+,15+,16-,20+/m0/s1

InChI key

FZALQGCQQJLGKE-BBNZOYGDSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjun Wu et al.
Molecular carcinogenesis, 56(10), 2190-2199 (2017-05-04)
Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) is a potent anti-tumor agent that triggers apoptosis in cells from multiple types of carcinoma but not in normal cells. However, diverse mechanisms are associated with insensitivity to TRAIL in various cancers. TRAIL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service