Skip to Content
MilliporeSigma
All Photos(1)

Documents

SIC006

Sigma-Aldrich

MISSION® siRNA Fluorescent Universal Negative Control #2, Cyanine 5

Synonym(s):

MISSION® siRNA Fluorescent Universal Negative Control, Cyanine 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

product line

MISSION®

color

Red (Cyanine 5)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Negative controls are an essential part of every siRNA experiment, and here we offer the siRNA Universal Negative Control #2 conjugated to Cyanine 5 for easy visualization and assessment of transfection efficiency.

Features and Benefits

  • Cyanine 5 labeled siRNA Universal Negative Control allows easy visualization of the siRNA in your cells by using fluorescence microscopy.
  • Rapidly assess transfection efficiency.
  • Ready to use.
  • Available in three sizes for your convenience.
  • Compatible with Human, Mouse, and Rat cells.

Legal Information

All siRNA oligos are sold under license from the Massachusetts Institute of Technology.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service