Skip to Content
MilliporeSigma
All Photos(1)

Documents

SHC015V

Sigma-Aldrich

MISSION® pLKO.1-puro-UbC-TagFP635 Positive Control Transduction Particles

Contains a gene encoding TagFP635, under the control of the UbC promoter

Synonym(s):

MISSION®, MISSION® Control Transduction Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

MISSION®

technique(s)

capture ELISA: 106 TU/mL using p24

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. In addition, the Control Transduction Particles are pseudotyped with an envelope G glycoprotein from Vesicular Stomatitis Virus (VSV-G), allowing transduction of a wide variety of mammalian cells. 200 μl of 106 TU/ml (via p24 titering assay) lentiviral particles are provided as frozen stock.
When conducting experiments using MISSION® shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION Control Transduction Particles are a critical positive control to monitor transduction efficiency.
To see more application data, protocols, vector maps visit sigma.com/shrna.

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.
When conducting experiments using MISSION shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. Silencing of the CMV promoter can be a problem in some cell types. For these cells, the ubiquitin promoter (UbC) can be a viable alternative. The MISSION UbC-TagFP635 Control Transduction Particles are a positive control to monitor transduction efficiency for applications in which an alternate promoter is necessary. The MISSION UbC-TagFP635 Control Transduction Particles are produced from the sequence-verified 9226 base pair lentivirus plasmid, pLKO.1-puro-UbC-TagFP635 (SHC015).

This construct aids in interpretation of experimental design and results by providing a far-red fluorescent protein marker for monitoring success in transduction of your cells of interest.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
TagFP635 is a trademark of Evrogen Co.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service