Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SEP010

Millipore

Seppro® IgY14

Spin Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

capacity

1 mg total protein loading(14 μL of human plasma, average human protein concentration 70 mg/mL)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Seppro® IgY14 columns allow users to separate 14 highly abundant proteins (HAP) from plasma protein mixtures. The Avian polyclonal IgY (Immunoglobulin Yolk) antibodies, have a distinct, highly specific partitioning advantage over competing products, allowing the researcher to look at a much cleaner human protein mixture. Seppro® columns make further proteomics analysis into the plasma proteome much easier and provide cleaner results.

Proteins Depleted:

Albumin
IgG
α1-Antitrypsin
IgA
IgM
Transferrin
Haptoglobin
α2-Macroglobulin
Fibrinogen
Complement C3
α1-Acid Glycoprotein (Orosomucoid)
HDL (Apolipoproteins A-I and A-II)
LDL (mainly Apolipoprotein B)

Application

Seppro® IgY14, spin columns (capacity: 15-20μl) is designed for use as a plasma/serum protein removal tool base upon immune affinity removal of 14 highly abundant proteins (HAP) from human plasma protein mixtures via liquid chromatography. Seppro® IgY14 is used to support the analysis of low abundance human serum/plasma proteins.[1][2]

Legal Information

Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • S4199Seppro® Dilution bufferSDS

  • S4324Seppro® Stripping BufferSDS

  • S4449Seppro® Neutralization BufferSDS

  • CLS8163Corning® Costar® Spin-X® centrifuge tube filters, cellulose acetate membrane, pore size 0.45 μm, non-sterileSDS

  • S4574Spin Column for Seppro®SDS

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hoi Pang Low et al.
Genomics, proteomics & bioinformatics, 11(6), 335-344 (2013-12-04)
Preeclampsia, a complication of pregnancy characterized by hypertension and proteinuria, has been found to reduce the subsequent risk for breast cancer in female offspring. As this protective effect could be due to exposure to preeclampsia-specific proteins during intrauterine life, the
Maneesh Bhargava et al.
PloS one, 9(10), e109713-e109713 (2014-10-08)
Acute Respiratory Distress Syndrome (ARDS) continues to have a high mortality. Currently, there are no biomarkers that provide reliable prognostic information to guide clinical management or stratify risk among clinical trial participants. The objective of this study was to probe
Inmaculada Lopez-Font et al.
Molecular neurobiology, 56(12), 8603-8616 (2019-07-11)
The β-site amyloid precursor protein cleaving enzyme 1 (BACE1) is the main brain β-secretase responsible for the amyloidogenic processing of the amyloid precursor protein (APP). Previous studies have suggested that cerebrospinal fluid (CSF) β-secretase activity may be a candidate diagnostic
Chien-Lun Chen et al.
Journal of proteomics, 85, 28-43 (2013-05-02)
In this study, we evaluated the reproducibility of abundant urine protein depletion by hexapeptide-based library beads and an antibody-based affinity column using the iTRAQ technique. The antibody-based affinity-depletion approach, which proved superior, was then applied in conjunction with iTRAQ to
Renato Millioni et al.
PloS one, 6(5), e19603-e19603 (2011-05-17)
To date, the complexity of the plasma proteome exceeds the analytical capacity of conventional approaches to isolate lower abundance proteins that may prove to be informative biomarkers. Only complex multistep separation strategies have been able to detect a substantial number

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service