Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0238

Sigma-Aldrich

Substance P Peptide

≥95% (HPLC), lyophilized, Succinyl-Asp⁶,Me-Phe⁸

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H55N7O11S1
Molecular Weight:
841.97
UNSPSC Code:
12352209
NACRES:
NA.32

product name

[Succinyl-Asp6,Me-Phe8]-Senktide Substance P 6-11,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Suc-Asp-Phe-NMe-Phe-Gly-Leu-Met-NH2

Application

Senktide (succinyl-[Asp6,Me-Phe8] substance P (6-11)), a substance P analogue, is used as a selective agonist of tachykinin NK-3 (or SP-N) receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service