Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCP0237

Sigma-Aldrich

PAR-1 Selective Agonist

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H53N9O6
Molecular Weight:
647.81
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Thr-Phe-Leu-Leu-Arg-NH2

Application

PAR-1 selective agonist has been used for platelet aggregation.
Thr-Phe-Leu-Leu-Arg-NH2 (TFLLR-NH2) is used as a selective agonist for protease-activated receptor 1 (PAR1) which is involved in the development of cardiac remodeling and hypertrophy after myocardial injury.

Biochem/physiol Actions

Protease-activated receptor-1 (PAR-1) is a G protein-coupled receptor, that is activated through thrombin and other proteases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rac Regulates the TRAP-Induced Release of Phosphorylated-HSP27 from Human Platelets via p38 MAP Kinase but Not JNK
Uematsu K, et al.
Cellular Physiology and Biochemistry, 49(4), 1523-1538 (2018)
High-resolution crystal structure of human protease-activated receptor 1
Zhang C, et al.
Nature, 492(7429), 387-387 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service