Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0183

Sigma-Aldrich

LH-RH II alligator, chicken

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H69N17O13
Molecular Weight:
1236.30
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Glp-His-Trp-Ser-His-Gly-Trp-Tyr-Pro-Gly-NH2

Application

LH-RH II alligator, chicken is used as a comparative LHRH in studies of LHRH evolution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacia A Sower et al.
General and comparative endocrinology, 161(1), 20-29 (2008-12-17)
The acquisition of a hypothalamic-pituitary axis was a seminal event in vertebrate evolution leading to the neuroendocrine control of many complex functions including growth, reproduction, osmoregulation, stress and metabolism. Lampreys as basal vertebrates are the earliest evolved vertebrates for which
Masafumi Amano et al.
Zoological science, 38(1), 51-59 (2021-03-01)
In vertebrates, gonadotropin-releasing hormone (GnRH) regulates gonadal maturation by stimulating the synthesis and release of pituitary gonadotropins. GnRH has also been identified in invertebrates. Crustacea consists of several classes including Cephalocarida, Remipedia, Branchiopoda (e.g., tadpole shrimp), Hexanauplia (e.g., barnacle) and
Scott I Kavanaugh et al.
Endocrinology, 149(8), 3860-3869 (2008-04-26)
We cloned a cDNA encoding a novel (GnRH), named lamprey GnRH-II, from the sea lamprey, a basal vertebrate. The deduced amino acid sequence of the newly identified lamprey GnRH-II is QHWSHGWFPG. The architecture of the precursor is similar to that
Stacia A Sower et al.
General and comparative endocrinology, 148(1), 22-32 (2006-01-24)
We have identified the first and perhaps only gonadotropin beta-like protein by cDNA cloning in sea lamprey, a member of the oldest lineage of vertebrates, the agnathans. Two pituitary gonadotropins (GTHs: follicle-stimulating hormone (FSH) and luteinizing hormone (LH)) have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service