Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0104

Sigma-Aldrich

Caspase Substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H41N7O11
Molecular Weight:
711.72
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Leu-Glu-His-Asp-AMC

Application

Ac-LEHD-AMC is used as a fluorogenic substrate of caspase 3 and caspase 9.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul M Kasili et al.
Journal of the American Chemical Society, 126(9), 2799-2806 (2004-03-05)
We demonstrate for the first time, the application and utility of a unique optical sensor having a nanoprobe for monitoring the onset of the mitochondrial pathway of apoptosis in a single living cell by detecting enzymatic activities of caspase-9. Minimally
M Nagase et al.
Bioscience, biotechnology, and biochemistry, 65(8), 1741-1747 (2001-10-02)
The molecules participating in apoptosis induced by T-2 toxin in human leukemia HL-60 cells were investigated. The rank order of the potency of trichothecene mycotoxins to induce internucleosomal DNA fragmentation was found to be T-2, satratoxin G, roridin A >
Masahiro Nagase et al.
Immunology letters, 84(1), 23-27 (2002-08-06)
Satratoxins have been recognized as potential immunomodulatory agents in outbreaks of building-related illness. Here we report that satratoxin G-treated human leukemia HL-60 cells underwent apoptosis through the action of caspase-3 which was activated by both caspase-8 and caspase-9. Western blot
Ikue Suganuma et al.
Insect biochemistry and molecular biology, 41(11), 909-921 (2011-09-14)
We cloned and characterized a novel Bombyx mori homologue (bm-dronc) of Drosophila melanogaster dronc (dm-dronc), which could encode a polypeptide of 438 amino acid residues. Bm-Dronc shares relatively low amino acid sequence identities of 25% and 26% with Dm-Dronc and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service