Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0059

Sigma-Aldrich

Calcitonin Acetyl salmon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C147H242N44O49S2
Molecular Weight:
3473.89
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2

Application

Calcitonin, a 32 amino acid polypeptide, is involved in the regulation of calcium and phosphorous metabolism often antagonistic to parathyroid hormone (PTH).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and evaluation of bone targeting salmon calcitonin analogs in normal and osteoporotic rats.
Bhandari KH, Newa M, Chapman J, et al.
J. Controlled Release, 158, 44-52 (2012)
Claudia Mueller et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 79(3), 646-657 (2011-06-28)
Protein aggregation, which is triggered by various factors, is still one of the most prevalent problems encountered during all stages of protein formulation development. In this publication, we present novel excipients, tryptophan-mPEGs (Trp-mPEGs) of 2 and 5 kDa molecular weight
Aiwen Huang et al.
Drug delivery, 18(8), 562-569 (2011-08-10)
Therapeutic peptide and protein drugs have high specificity and activity in their functions but present challenges in their administration route, requiring development of new delivery systems to improve their bioavailability. The aim of this work was to investigate the role
Cetin Tas et al.
International journal of pharmaceutics, 423(2), 257-263 (2011-12-17)
Peptides and polypeptides have important pharmacological properties but only a limited number have been exploited as therapeutics because of problems related to their delivery. Most of these drugs require a parenteral delivery system which introduces the problems of pain, possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service