Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0023

Sigma-Aldrich

ANP 126-150 rat /Auriculin B, ANP 4-28 rat

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C113H179N39O35S2
Molecular Weight:
2708.00
UNSPSC Code:
51111800
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr

Application

Atrial natriuretic peptide(s) (ANP) (Atriopeptin) are peptide hormones that regulate vasodilation. The ANP fragment Atriopeptin III (ANP 126-150) is used to study the regulation of Na+/H+ exchange mediated in erythrocytes by cGMP/guanylate-cyclase activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Nortier et al.
Peptides, 14(2), 405-408 (1993-03-01)
Incubation of rANP(5-28)--also called atriopeptin III (AP III)--with purified endopeptidase 24.11 led preferentially to the production of Phe-Arg-Tyr, while other products of minor importance were detected. One of these was identified as rANP(5-25) (atriopeptin I) (AP I). This hydrolysis pattern
B M Schmitt et al.
Clinical and experimental pharmacology & physiology, 24(2), 131-138 (1997-02-01)
1. In mammalian plasma, many different inhibitors of Na+/K(+)-ATPase are present, but it is not clear whether their net effect on NA+/K(+)-ATPase activity changes during the regulation of electrolyte and fluid balance. We studied Na+/K(+)-ATPase inhibition by plasma extracts in
V Petrov et al.
European journal of biochemistry, 221(1), 195-199 (1994-04-01)
Human atrial natriuretic peptide (ANP) fragments ANP-(127-150) or ANP-III and ANP-(127-149) or ANP-II activate Na+/H+ exchange in human erythrocytes at concentrations as low as 1 pM. Both ANP-(127-147) or ANP-I and ANP-(129-150) or des-Ser5, Ser6-ANP-III have no effect on erythrocyte
V Petrov et al.
The American journal of physiology, 271(5 Pt 1), C1556-C1564 (1996-11-01)
Guanylate cyclase activity in human erythrocytes is investigated by evaluating the intracellular guanosine 3',5'-cyclic monophosphate (cGMP) content in the presence of various agents that exert specific effects on soluble or particulate guanylate cyclase. The increase in the intraerythrocyte cGMP content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service