Skip to Content
MilliporeSigma
All Photos(1)

Documents

SBVT27

Sigma-Aldrich

SB-cyBSEP-HEK293

Synonym(s):

Solvo membrane vesicles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

recombinant

expressed in HEK 293 cells

form

liquid

packaging

pkg of 500 μL

concentration

5 mg/mL

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Membrane Preparations for Vesicular Transport Assays (VT) are suitable for general drug-efflux transporter interaction studies. Both substrate and inhibitor interactions can be assessed using vesicles. The success of substrate interaction studies strongly depends on the passive permeability of the compound. High permeability substrates might not be detected. Control Membranes with no, or significantly lower transporter activity are also available.

Application

In the vesicular transport assay so-called "inside-out" membrane vesicles containing ABC transporters are applied. Incubating substrates of the respective efflux transporter in the presence of the inverted membrane vesicles and ATP will allow measuring accumulation of the substrates into the vesicles. In many cases radiolabeled reporter substrates are used but recently Solvo developed the new PREDIVEZ Vesicular Transport Kits that use fluorescent reporter substrates.

The standard vesicular transport assay is an inhibitory assay performed with cold test articles. This assay provides information on any interaction between the ABC transporter and the test article. The transport of the reporter substrate is measured in the presence of the test article (typically in 7 concentrations) and IC50 is defined as the concentration inhibiting the transport of the reporter substrate by 50%.

Should radiolabeled form of the investigated compound or adequate analytical methods (LC/MS, HPLC) be available, the vesicular transport assay may be performed in a direct format without the reporter substrate and may identify substrate nature of the test article. The vesicular transport substrate assay is a low throughput assay. It is suitable for low permeability test compounds as high permeability compounds may escape from the vesicles through the lipid bilayer.

Physical form

Supplied as frozen membrane vesicles, containing 5 mg/mL membrane protein, labeled with volume, catalog number (transporter) and date of production.

Legal Information

PREDIVEZ is a trademark of Solvo Biotechnology

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service