Skip to Content
MilliporeSigma
All Photos(1)

Documents

SBR00094

Sigma-Aldrich

Valnemulin hydrochloride

new

≥96% (HPLC), powder

Synonym(s):

Econor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52N2O5S · HCl
CAS Number:
Molecular Weight:
601.28
MDL number:
UNSPSC Code:
51101500

Quality Level

Assay

≥96% (HPLC)

form

powder

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | inhibits

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(C)[C@@H](N)C(NCC(C)(C)SCC(O[C@H]1[C@](C)([C@@H]2C)[C@@]3([H])[C@@](CC2)(CCC3=O)[C@@H](C)[C@H](O)[C@](C)(C=C)C1)=O)=O.Cl

InChI

1S/C31H52N2O5S.ClH/c1-10-29(8)15-22(38-23(35)16-39-28(6,7)17-33-27(37)24(32)18(2)3)30(9)19(4)11-13-31(20(5)26(29)36)14-12-21(34)25(30)31;/h10,18-20,22,24-26,36H,1,11-17,32H2,2-9H3,(H,33,37);1H/t19-,20+,22-,24-,25+,26+,29-,30+,31+;/m1./s1

InChI key

MFBPRQKHDIVLOJ-AFFLPQGKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Valnemulin Hydrochloride is an antibiotic which belongs to the class of compounds known as pleuromutilins. It is a semisynthetic derivative of the naturally occurring antibiotic tiamulin. Valnemulin Hydrochloride has activity against various Gram-positive and some Gram-negative bacteria, including Mycoplasma and some anaerobic bacteria, as well as the protozoan parasite Eimeria.

Biochem/physiol Actions

Valnemulin Hydrochloride inhibits bacterial protein synthesis by binding to the 50S subunit of the bacterial ribosome. More specifically, it binds to the peptidyl transferase center of the ribosome and interferes with the transfer of amino acids to growing polypeptide chains, leading to the inhibition of protein synthesis and bacterial death.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service