Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SBR00074

Sigma-Aldrich

Asunaprevir

≥99% (HPLC), powder

Synonym(s):

SM-5887, BMS650032, 3-Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}-L-valyl-(4R)-4-[(7-chloro-4-methoxy-1-isoquinolinyl)oxy]-N-{(1R,2S)-1-[(cyclopropylsulfonyl)carbamoyl]-2-vinylcyclopropyl}-L-prolinamide, ASV, BMS 650032, BMS-650032

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H46ClN5O9S
CAS Number:
Molecular Weight:
748.29
MDL number:
UNSPSC Code:
51101500
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

powder

color

white to off-white

antibiotic activity spectrum

viruses

Mode of action

protein synthesis | inhibits

storage temp.

−20°C (−15°C to −25°C)

SMILES string

CC(C)(C)[C@@H](C(=O)N1C[C@@H](C[C@H]1C(=O)N[C@@]2(C[C@H]2C=C)C(=O)NS(=O)(=O)C3CC3)OC4=NC=C(C5=C4C=C(C=C5)Cl)OC)NC(=O)OC(C)(C)C

InChI

1S/C35H46ClN5O9S/c1-9-19-16-35(19,31(44)40-51(46,47)22-11-12-22)39-28(42)25-15-21(49-29-24-14-20(36)10-13-23(24)26(48-8)17-37-29)18-41(25)30(43)27(33(2,3)4)38-32(45)50-34(5,6)7/h9-10,13-14,17,19,21-22,25,27H,1,11-12,15-16,18H2,2-8H3,(H,38,45)(H

InChI key

XRWSZZJLZRKHHD-QGQNBIQGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Asunaprevir is a competitive inhibitor of HCV NS3/4A protease which act by inhibiting the protease activity of the NS3/4A enzyme of the hepatitis C virus.

Biochem/physiol Actions

Asunaprevir is a direct acting antiviral agent that inhibits the NS3/4A protease of the hepatitis C virus (HCV). Inhibition of the NS3/4A protease prevents viral replication and leads to the clearance of HCV from the body. Asunaprevir is active again viruses.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service