Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SBCT03

Sigma-Aldrich

SB beta gal Sf9 CTRL

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

Negative control Sf9 insect membrane, recombinant, expressed in baculovirus infected Sf9 cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$220.00

$220.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$220.00

About This Item

UNSPSC Code:
12352202

$220.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

form

liquid

concentration

5.0 mg/mL

color

off-white

shipped in

dry ice

storage temp.

−70°C

General description

Membrane Preparations for Vesicular Transport Assays (VT) are suitable for general drug-efflux transporter interaction studies. Both substrate and inhibitor interactions can be assessed using vesicles. Membrane Preparations for ATPase Assays are suitable for drug-efflux transporter interaction studies based on ATPase activity determination, and could be used for differentiation between transporter substrates and inhibitors. Control Membranes with no-, or significantly lower transporter activity are available for both Vesicular Transport and ATPase Assays.

Application

Control Membranes with no-, or significantly lower transporter activity provide appropriate negative controls for both Vesicular Transport and ATPase assays. Refer to individual Vesicular Transport or ATPase assay product detail pages for appropriate control membranes needed for a specific assay.

Packaging

250 μL

Physical form

Supplied as frozen membrane vesicles, containing 5 mg/ml membrane protein, labeled with volume, catalog number (transporter) and date of production.

Legal Information

Distributed for SOLVO Biotechnology, Inc

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service