Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAE0094

Sigma-Aldrich

SAvPhire Monomeric Streptavidin

recombinant, expressed in E. coli, High Affinity

Synonym(s):

mSA3

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$398.00
5 MG
$1,150.00

$398.00


Available to ship onApril 17, 2025Details



Select a Size

Change View
1 MG
$398.00
5 MG
$1,150.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

$398.00


Available to ship onApril 17, 2025Details


recombinant

expressed in E. coli

form

lyophilized powder

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

SAvPhire Monomeric Streptavidin is a variant of the biotin-binding streptavidin, which naturally exists as a tetramer. Many monomeric streptavidin proteins have rapid dissociation kinetics, which prevent their use in stable labeling applications. However, SAvphire Monomeric Streptavidin has been engineered to have a dissociation rate comparable to that of tetrameric streptavidin.
The size and monovalency of this product provide advantages over tetrameric streptavidin in applications such as functional cell surface labeling and atomic force spectroscopy. These advantages include:
  • Minimizing distribution or dynamics in the probing of biotinylated targets
  • Reducing risk of aggregation, precipitation, and cross-linking in pre-incubation steps with biotinylated molecules
  • Easier access to desired target

This product can be used in traditional biotin-binding applications, including Western blotting, immunoprecipitation, immunohistochemistry, and flow cytometry.

Physical properties

Sequence contains N-terminal 6×-Histidine-tag and a C-terminal FLAG-tag

Reconstitution

Once reconstituted, the solution can be stored at 2-8 °C for at least 1 month. For long term storage, aliquot and store at -20 °C.
This product is lyophilized from PBS, pH 7.4. Reconstitute the vial contents to a 4 mg/mL solution by adding water as follows:

  • 1 mg vial: add 250 μL of water for a 4 mg/mL solution
  • 5 mg vial: add 1.25 mL of water for a 4 mg/mL solution

The reconstituted product can be further diluted in water or PBS as needed.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
SAvPhire is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and purification of soluble monomeric streptavidin in Escherichia coli
Demonte D, et al.
Applied Microbiology and Biotechnology, 98(14), 6285-6295 (2014)
Functional expression of monomeric streptavidin and fusion proteins in Escherichia coli: applications in flow cytometry and ELISA.
Kroetsch, A. et al.
Applied Microbiology and Biotechnology, 102, 10079-10089 (2018)

Questions

1–2 of 2 Questions  
  1. Could you provide the molar extinction coefficient for this product?

    1 answer
    1. The extinction coefficient for the product in question is 2.8 (E0.1%).

      Helpful?

  2. What is the isoelectric point (pI) of Product No. SAE0094, SAvPhire™ Monomeric Streptavidin?

    1 answer
    1. The isoelectric point (pI) of Product No. SAE0094, SAvPhire™ Monomeric Streptavidin, is not determined by us. However, the theoretical pI is 5.9.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service