Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAE0065

Sigma-Aldrich

Lipase, produced by Aspergillus oryzae

>=100 LCLU/g

Synonym(s):

Eversa Transform 2.0

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Aspergillus sp. (A. oryzae)

Quality Level

form

liquid

density

~1.2 g/mL

shipped in

ambient

storage temp.

2-8°C

General description

This product is a liquid lipase product with high activity in the transesterifciation of glycerides, and in the esterification of free fatty acids (FFA). This product is useful for biodiesel applications, for the conversion of both glycerides and FFA into biodiesel, and for production of fatty acid methyl esters (FAME) in the presence of methanol. This enzyme works at 40 °C, which allows the processese to require less energy for cooling.

Legal Information

A product of Novozyme Corp

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of FAME and FAEE via Alcoholysis of Sunflower Oil by Eversa Lipases Immobilized on Hydrophobic Supports
Remonatto, D., et al.
Applied Biochemistry and Biotechnology (2018)
Andre Mong Jie Ng et al.
International journal of molecular sciences, 22(19) (2021-10-14)
Medium-chain triglycerides (MCTs) are an emerging choice to treat neurodegenerative disorders such as Alzheimer's disease. They are triesters of glycerol and three medium-chain fatty acids, such as capric (C8) and caprylic (C10) acids. The availability of C8-C10 methyl esters (C8-C10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service