Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB6010057

Sigma-Aldrich

Anti-phospho-ERN1 (S724) antibody, clone 1F12, Rabbit Monoclonal

new

recombinant, expressed in HEK293F cells

Synonym(s):

Serine/threonine-protein kinase/endoribonuclease IRE1, Endoplasmic reticulum-to-nucleus signaling 1, hIRE1p, Ire1-alpha, IRE1a, Serine/threonine-protein kinase, Endoribonuclease, ERN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

recombinant

expressed in HEK 293 F cells

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

1F12, recombinant monoclonal

form

liquid

mol wt

150 kDa

purified by

affinity chromatography

species reactivity

human

concentration

0.6mg/ml

technique(s)

ELISA: 2000:10000
flow cytometry: 1:50-1:200
immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

-10 to -25°C

target post-translational modification

phosphorylation (pSer724)

General description

Senses unfolded proteins in the lumen of the endoplasmic reticulum via its N-terminal domain which leads to enzyme auto-activation (PubMed:9637683, PubMed:11175748, PubMed:12637535). The active endoribonuclease domain splices XBP1 mRNA to generate a new C-terminus, converting it into a potent unfolded-protein response transcriptional activator and triggering growth arrest and apoptosis (PubMed:24508390).

Immunogen

A synthesized peptide derived from human Phospho-ERN1 (S724)

Sequence

GRH-pS-FSR

Physical form

Rabbit IgG in phosphate buffered saline , pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Storage and Stability

Upon receipt, store at -20°C or -80°C. Avoid repeated freeze.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service