Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB5801022

Sigma-Aldrich

Alpaca Anti-Mouse IgG1(Fcγ) HRP single domain Antibody

recombinant Alpaca(VHH) Nano antibody

Synonym(s):

Recombinant Alpaca single variable domain secondary Nanobody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

alpaca

recombinant

expressed in E. coli

conjugate

horse radish peroxidase (HRP)

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

recombinant monoclonal

form

lyophilized solid

mol wt

14 kDa

species reactivity

mouse, Alpaca

technique(s)

ELISA: 1:10000-1:50000
western blot: 1:10000-1:50000

size

0.1 mL

color

colorless

isotype

VHH

shipped in

wet ice

storage temp.

2-8°C

General description

Alpaca Anti-Mouse IgG1(Fcγ) HRP single domain Antibody is designed to specifically detect the Fc region of mouse IgG1. It is a monovalent, single domain, recombinant antibody to mouse IgG1(Fcγ fragment specific) coupled to HRP. Based on immunoelectrophoresis and/or ELISA, Alpaca Anti-Mouse IgG1(Fcγ) HRP single domain Antibody detects the Fc region of mouse IgG1 selectively but not with other mouse IgG subclasses, mouse IgM, or the Fab portion of mouse immunoglobulins.


Most monoclonal antibodies are generated in mouse. There are five antibody isotypes (IgA, IgD, IgE, IgG, and IgM) from mouse. Each isotype has a different heavy chain. Mouse IgG constitutes 75% of serum immunoglobulins, and IgG is the predominant form of first antibody produced from mouse. Mouse IgG consists of five subclasses-IgG1, IgG2a, IgG2b, IgG2c(inbred mouse strains with the Igh1-b allele have IgG2c isotype instead of IgG2a), IgG3. They are highly homologous and differ mainly in the hinge region. The whole IgG molecule possesses both the Fc region and the Fab region, which possessing the epitope-recognition site. The IgG contains two heavy and light chains, and the heavy chain is about 50 KD and the light chain is about 25 KD. The common IgG is monomeric with a molecular weight of approximately 150 kD.

VHH are single-domain antibodies derived from the variable regions of heavy chain of Camelidae immunoglobulin. The size of VHH is extremely small(less than15KDa) compared to other forms of antibody fragment, which significantly increase the permeability of VHH. Thus VHH is considered of great value for research, diagnostics and therapeutics.

Specificity

Mouse IgG1(Fcγ fragment specific)
Isotype: VHH domain of alpaca IgG2b/2c

Immunogen

Recombinant Alpaca single variable domain secondary nano antibody

Packaging

100 μg in Glass bottle

Physical form

Lyophilized from 10mM PBS (pH 7.5), 0.05% sucrose, 0.1% trehalose

Reconstitution

Add 100uL dH2O or 100uL dH2O with 50% Glycerol to a final concentration of 1mg/mL

Storage and Stability

After reconstitution, store at –20 °C(Avoid freeze / thaw cycles). Protect from light. Stable for 12 months at -20°C

Other Notes

It has weak cross reaction with mouse IgG3. No cross-reactivity with mouse IgG2b/2c, mouse IgM, rabbit, human, cynomolgus, rat, goat IgG

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service