Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5703016

Sigma-Aldrich

Anti-Thioredoxin reductase 2 (TXNRD2 ) Antibody, clone 4O1K0, Rabbit Monoclonal

Synonym(s):

SELZ, TR, TR-BETA, TR3, TRXR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

material

colorless

clone

4O1K0, monoclonal

form

liquid

mol wt

56 kDa

species reactivity

rat, human, rat

concentration

0.79 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MAAMAVALRGLGGRFRWRTQAVAGGVRGAARGAAAGQRDYDLLVVGGGSGGLACAKEAAQLGRKVAVVDYVEPSPQGTRWGLGGTCVNVGCIPKKLMHQA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TXNRD2(10587)

General description

This gene encodes a member of the class I pyridine nucleotide-disulfide oxidoreductase family. The encoded protein is a selenocysteine-containing flavoenzyme that maintains thioredoxins in a reduced state, thereby playing a key role in regulating the cellular redox environment. Mammals have three related thioredoxin reductases. This gene encodes a mitochondrial form important for scavenging of reactive oxygen species in mitochondria. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq, Sep 2013]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Thioredoxin reductase 2 (TXNRD2 ) (Q9NNW7).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service