Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

SAB5702943

Sigma-Aldrich

Anti-Syntaxin Antibody, clone 7D9V8, Rabbit Monoclonal

Synonym(s):

GEFSP9, STX1B1, STX1B2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$533.00

$533.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$533.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$533.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

7D9V8, monoclonal

form

liquid

mol wt

35 kDa

species reactivity

human, rat, rat

concentration

0.65 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MKDRTQELRSAKDSDDEEEVVHVDRDHFMDEFFEQVEEIRGCIEKLSEDVEQVKKQHSAILAAPNPDEKTKQELEDLTADIKKTANKVRSKLKAIEQSIE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STX1B(112755)

General description

The protein encoded by this gene belongs to a family of proteins thought to play a role in the exocytosis of synaptic vesicles. Vesicle exocytosis releases vesicular contents and is important to various cellular functions. For instance, the secretion of transmitters from neurons plays an important role in synaptic transmission. After exocytosis, the membrane and proteins from the vesicle are retrieved from the plasma membrane through the process of endocytosis. Mutations in this gene have been identified as one cause of fever-associated epilepsy syndromes. A possible link between this gene and Parkinson's disease has also been suggested. [provided by RefSeq, Jan 2015]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Syntaxin (P61266).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service