Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702905

Sigma-Aldrich

Anti-Olig3 Antibody, clone 9C1N3, Rabbit Monoclonal

Synonym(s):

OLIG3, Bhlhb7, bHLHe20, oligodendrocyte transcription factor 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$533.00

$533.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$533.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$533.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

9C1N3, monoclonal

form

liquid

mol wt

29 kDa

species reactivity

human

concentration

0.50 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

HAANSVHPVHPILGGALSSGNASSPLSAASLPAIGTIRPPHSLLKAPSTPPALQLGSGFQHWAGLPCPCTICQMPPPPHLSALSTANMARLSAESKDLLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... Olig3(167826)

General description

May determine the distinct specification program of class A neurons in the dorsal part of the spinal cord and suppress specification of class B neurons.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 173-272 of human Olig3 (Q7RTU3).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service