Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5702888

Sigma-Aldrich

Anti-PDK1/PDHK1 Antibody, clone 5T6S8, Rabbit Monoclonal

Synonym(s):

PDK1, pyruvate dehydrogenase kinase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$533.00

$533.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$533.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$533.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

5T6S8, monoclonal

form

liquid

mol wt

49 kDa

species reactivity

rat, rat, human

concentration

1.00 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

STAPRPRVETSRAVPLAGFGYGLPISRLYAQYFQGDLKLYSLEGYGTDAVIYIKALSTDSIERLPVYNKAAWKHYNTNHEADDWCVPSREPKDMTTFRSA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDK1(5163)

General description

Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jun 2013]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 337-436 of human PDK1/PDHK1 (NP_002601.1).
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service