Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5702754

Sigma-Aldrich

Anti-AGL Antibody, clone 8V9K5, Rabbit Monoclonal

Synonym(s):

AGL, GDE, glycogen debranching enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

8V9K5, monoclonal

form

liquid

mol wt

175 kDa

species reactivity

rat, rat, human

concentration

1.20 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MGHSKQIRILLLNEMEKLEKTLFRLEQGYELQFRLGPTLQGKAVTVYTNYPFPGETFNREKFRSLDWENPTEREDDSDKYCKLNLQQSGSFQYYFLQGNE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGL(178)

General description

This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human AGL (P35573).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service