Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702713

Sigma-Aldrich

Anti-p150 CAF1 Antibody, clone 1T1O5, Rabbit Monoclonal

Synonym(s):

CAF-1, CAF1, CAF1B, CAF1P150, P150

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

1T1O5, monoclonal

form

liquid

mol wt

150 kDa

species reactivity

human

concentration

0.14mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

NFLRNRIETSIGQSTVIIDLTEDSNEQPDSLVDHNKLNSEASPSREAINGQREDTGDQQGLLKAIQNDKLAFPGETLSDIPCKTEEEGVGCGGAGRRGDSQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHAF1A(10036)

General description

Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B; MIM 601245), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro (Kaufman et al., 1995 [PubMed 7600578]).[supplied by OMIM, Mar 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 100-200 of human p150 CAF1 (Q13111).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service