Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702651

Sigma-Aldrich

Anti-BST2 Antibody, clone 4L6O2, Rabbit Monoclonal

Synonym(s):

CD317, TETHERIN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

4L6O2, monoclonal

form

liquid

mol wt

35 kDa

species reactivity

human, rat, rat

concentration

0.37 mg/mL

technique(s)

immunofluorescence: 1:50-1:500
immunohistochemistry: 1:50-1:500
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

LQQELTEAQKGFQDVEAQAATCNHTVMALMASLDAEKAQGQKKVEELEGEITTLNHKLQDASAEVERLRRENQVLSVRIAD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BST2(684)

General description

Bone marrow stromal cells are involved in the growth and development of B-cells. The specific function of the protein encoded by the bone marrow stromal cell antigen 2 is undetermined; however, this protein may play a role in pre-B-cell growth and in rheumatoid arthritis. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 70-150 of human BST2 (Q10589).
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IF/ICC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service