Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702600

Sigma-Aldrich

Anti-PHOX2B Antibody, clone 4W5J8, Rabbit Monoclonal

Synonym(s):

NBLST2, NBPhox, PMX2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

4W5J8, monoclonal

form

liquid

mol wt

35 kDa

species reactivity

mouse, mouse

concentration

0.55 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

GGGPSPAGAPGAAGPGGPGGEPGKGGAAAAAAAAAAAAAAAAAAAAGGLAAAGGPGQGWAPGPGPITSIPDSLGGPFASVLSSLQRPNGAKAALVKSSMF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHOX2B(8929)

General description

The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element. Expansion of a 20 amino acid polyalanine tract in this protein by 5-13 aa has been associated with congenital central hypoventilation syndrome. [provided by RefSeq, Jul 2016]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 215-314 of human PHOX2B (Q99453).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service