Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5702577

Sigma-Aldrich

Anti-TSC1 Antibody, clone 6Q10T10, Rabbit Monoclonal

Synonym(s):

LAM, TSC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

6Q10T10, monoclonal

form

liquid

mol wt

150 kDa

species reactivity

human

concentration

0.49 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

TTMGEASASIPTTVGSLPSSKSFLGMKARELFRNKSESQCDEDGMTSSLSESLKTELGKDLGVEAKIPLNLDGPHPSPPTPDSVGQLHIMDYNETHHEHS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSC1(7248)

General description

This gene encodes a growth inhibitory protein thought to play a role in the stabilization of tuberin. Mutations in this gene have been associated with tuberous sclerosis. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jun 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1065-1164 of human Hamartin (Q92574).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service