Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB5702503

Sigma-Aldrich

Anti-HAPLN1 Antibody, clone 5O9U2, Rabbit Monoclonal

Synonym(s):

CRT1, CRTL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

5O9U2, monoclonal

form

liquid

mol wt

57 kDa

species reactivity

rat, rat, human

concentration

2.2 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

VFCFTSNFNGRFYYLIHPTKLTYDEAVQACLNDGAQIAKVGQIFAAWKILGYDRCDAGWLADGSVRYPISRPRRRCSPTEAAVRFVGFPDKKHKLYGVYCFRAYN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HAPLN1(1404)

General description

Stabilizes the aggregates of proteoglycan monomers with hyaluronic acid in the extracellular cartilage matrix.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 250-354 of human HAPLN1 (P10915).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service