Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5702307

Sigma-Aldrich

Anti-MSR1 Antibody, clone 10O8B0, Rabbit Monoclonal

Synonym(s):

CD204, SCARA1, SR-A, SR-AI, SR-AII, SR-AIII, SRA, phSR1, phSR2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$533.00

$533.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$533.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$533.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

10O8B0, monoclonal

form

liquid

mol wt

70 kDa

species reactivity

rat, rat, human

concentration

1.40 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MEQWDHFHNQQEDTDSCSESVKFDARSMTALLPPNPKNSPSLQEKLKSFKAALIALYLLVFAVLIPLIGIVAAQLLKWETKNCSVSSTNANDITQSLTGK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MSR1(4481)

General description

This gene encodes the class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing of this gene. These receptors or isoforms are macrophage-specific trimeric integral membrane glycoproteins and have been implicated in many macrophage-associated physiological and pathological processes including atherosclerosis, Alzheimer's disease, and host defense. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. It has an altered intracellular processing and is trapped within the endoplasmic reticulum, making it unable to perform endocytosis. The isoform type 3 can inhibit the function of isoforms type 1 and type 2 when co-expressed, indicating a dominant negative effect and suggesting a mechanism for regulation of scavenger receptor activity in macrophages. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MSR1 (P21757).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service