This product is a SARS-CoV-2 Spike S2 ECD protein.
SAB5700597
Recombinant 2019-nCoV Spike S2 ECD Protein with His tag
Synonym(s):
S glycoprotein Subunit2, S2 protein, Spike glycoprotein Subunit2, Spike protein S2
Select a Size
$611.00
Select a Size
About This Item
$611.00
Recommended Products
recombinant
expressed in baculovirus infected insect cells
Quality Level
Assay
>90% (SDS-PAGE)
form
lyophilized powder
mol wt
58.8 kDa
impurities
<1.0 EU/μg (LAL test)
<1.0 EU/μg protein (LAL test)
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
SARS virus ... 3C(43740568)
General description
Reconstitution
Storage and Stability
Disclaimer
Data presented is the available current product information and provided as-is. These products have not been tested or verified in any additional applications, sample types, including any clinical use. Experimental conditions must be empirically derived by the user. Our Antibody Guarantee only covers tested applications stated herein and conditions presented in our product information and is not extended to publications.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is it from SARS CoV 2 or SARS CoV 1?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service