Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5700491

Sigma-Aldrich

Anti-Phospho-PLCG1-S1248 antibody produced in rabbit

Synonym(s):

NCKAP3, PLC-II, PLC1, PLC148, PLCG1, PLCgamma1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$437.00

$437.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$437.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$437.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

148

species reactivity

mouse, human, rat

concentration

1.08 mg/ml

technique(s)

immunohistochemistry: 1:10-1:100
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer1248)

Gene Information

human ... PLCG1(5335)

General description

The protein encoded by this gene catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of receptor-mediated tyrosine kinase activators. For example, when activated by SRC, the encoded protein causes the Ras guanine nucleotide exchange factor RasGRP1 to translocate to the Golgi, where it activates Ras. Also, this protein has been shown to be a major substrate for heparin-binding growth factor 1 (acidic fibroblast growth factor)-activated tyrosine kinase. Two transcript variants encoding different isoforms have been found for this gene.

Immunogen

A synthetic phosphorylated peptide around S1248 of human PLCG1 (NP_002651.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service