Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5700084

Sigma-Aldrich

Anti-KRAS Antibody

rabbit polyclonal

Synonym(s):

C-K-RAS, CFC2, K-RAS2A, K-RAS2B, K-RAS4A, K-RAS4B, KI-RAS, KRAS, KRAS1, KRAS2, NS, NS3, RALD, RASK2, c-Ki-ras2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$583.00

$583.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$583.00


Please contact Customer Service for Availability

Product Name

Anti-KRAS antibody produced in rabbit,

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21

species reactivity

rat, mouse, human

concentration

3.23 mg/ml

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRAS(3845)

General description

This gene, a Kirsten ras oncogene homolog from the mammalian ras gene family, encodes a protein that is a member of the small GTPase superfamily. A single amino acid substitution is responsible for an activating mutation. The transforming protein that results is implicated in various malignancies, including lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas and colorectal carcinoma. Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region.

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 30-188 of human KRAS (NP_004976.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service