Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5500051

Sigma-Aldrich

Anti-CD21 antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP199, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP199, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

mouse

technique(s)

flow cytometry: 1:100
immunoblotting: 1:150
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CR2(1380)

General description

CD21 antigen, also known as complement receptor type 2 (CR2), is a glycoprotein and a receptor for the C3d fragment of complement component 3. It is predominantly expressed in follicular dendritic cells and B lymphocytes. CD21 binds to CD19, and CD81 to form a B cell complex on mature B cells and enhances the B cell response to antigen through C3d.

Immunogen

Synthetic peptide derived from the C-terminus of human CD21 protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service