Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5200015

Sigma-Aldrich

Monoclonal Anti-Phosphotyrosine antibody produced in mouse

clone G104, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-Phosphotryrosine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product SAB5200015 is not currently sold in your country. Contact Technical Service

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

G104, monoclonal

form

buffered aqueous glycerol solution

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr)

Specificity

Reacts with phosphotyrosine, and detects the presence of phosphotyrosine in both un-stimulated and stimulated cell lysates. Does not cross-react with phosphoserine or phosphothreonine.

Immunogen

Phosphotyrosine, alanine and glyceine in a 1:1:1 ratio polymerized in the presence of keyhole limpet hemocyanin with 1-ethyl-3-(3′-dimentrylaminopropyl) carbodiimide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Yu et al.
Nucleic acids research, 49(10), 5779-5797 (2021-05-29)
Faithful genome integrity maintenance plays an essential role in cell survival. Here, we identify the RNA demethylase ALKBH5 as a key regulator that protects cells from DNA damage and apoptosis during reactive oxygen species (ROS)-induced stress. We find that ROS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service