Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4702104

Monoclonal Anti-CD271, purified produced in mouse

new

Synonym(s):

NGFR, TNFRSF16, p75NGFR, p75NTR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody product type

primary antibodies

clone

NGFR5, monoclonal

form

buffered aqueous solution

species reactivity

human, ( Non-human primates), ( Rabbit), ( Cat)

concentration

1 mg/ml

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NGFR(4804)

General description

CD271 / NGFR, also known as p75NGFR or p75NTR, is a 75 kDa low affinity receptor for the NGF (nerve growth factor), BDNF (brain-derived growth factor), and other neurotrophins, such as NT3 and NT4/5. Unlike other members of the tumor necrosis factor receptor superfamily of transmembrane proteins, CD271 has unique intracellular domain structure (lacks catalytic activity) and downstream signaling partners. Triggered by its ligands CD271 affects growth, differentiation, migration and death of the nervous system cells.

Specificity

The mouse monoclonal antibody NGFR5 (originally C34C) recognizes an epitope within ammino acids 1 - 160 of CD271/NGFR, a 75 kDa transmembrane glycoprotein of the TNFR superfamily.

Immunogen

Purified CD271 protein isolated from human melanoma cell line A875

Physical form

Phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified by protein-A affinity chromatography.

Storage and Stability

Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service