Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4702006

Monoclonal Anti-Cytokeratin 19 FITC produced in mouse

new

Synonym(s):

CK19, CYK19, K19, K1CS, KRT19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

FITC conjugate

Quality Level

antibody product type

primary antibodies

clone

A53-B/A2, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/ml

technique(s)

flow cytometry: suitable

isotype

IgG2a

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... KRT19(3880)

General description

Cytokeratins are a subfamily of intermediate filaments and characterized by remarkable biochemical diversity. They are represented in epithelial tissues by at least 20 different polypeptides, molecular weight between 40 kDa and 68 kDa. The individual cytokeratin polypeptides are designated 1 to 20 and divided into the type I (acidic cytokeratins 9-20) and type II (basic to neutral cytokeratins 1-8) families.

Specificity

The mouse monoclonal antibody A53-B/A2 recognizes the rod domain of cytokeratin 19 (40 kDa), an intracellular antigen constituting intermmediate cytoskeleton filaments. Cytokeratin 19 is not expressed in hepatocytes; it is often co-expressed with cytokeratin 7.

Immunogen

MCF-7 human breast adenocarcinoma cell line

Physical form

Phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with fluorescein isothiocyanate (FITC) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service