Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4702004

Monoclonal Anti-NG2 PE produced in mouse

new

Synonym(s):

CSPG4, Chondroitin sulfate proteoglycan 4, MCSP, MSK16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

7.1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CSPG4(1464)

General description

NG2 / chondroitin sulfate proteoglycan 4 is expressed on glial cell populations, but not on normal hepatopoietic cells. It is an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells, where it plays role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes, and supports signaling pathways important for tumor invasion and growth. NG2 also serves as an AML blast tumor marker associated with poor prognosis.

Specificity

The mouse monoclonal antibody 7.1 recognizes an extracellular epitope of NG2, the melanoma-associated chondroitin sulfate proteoglycan 4 of Mw approximately 220-300 kDa.

Immunogen

Human bone marrow stromal cells infected with SV-40

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service