Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700982

Sigma-Aldrich

Monoclonal Anti-PAG1 antibody produced in mouse

clone MEM-255

Synonym(s):

CBP, PAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
WB
clone:
MEM-255, monoclonal
technique(s):
western blot: 1 μg/mL
citations:

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-255, monoclonal

form

buffered aqueous solution

concentration

1 mg/mL

technique(s)

western blot: 1 μg/mL

accession no.

NM_018440.3

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PAG1(55824)

General description

The antibody MEM-255 recognizes an epitope (aa 235-280) of Csk-binding protein (Cbp) located in the cytoplasmic domain, also known as protein associated with glycosphingolipid-enriched microdomains (PAG).

Immunogen

Recombinant intracellular fragment (aa 97-432) of human Cbp (PAG).

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service