Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700639

Sigma-Aldrich

Monoclonal Anti-HLA Class I-FITC antibody produced in mouse

clone W6/32, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$433.00

$433.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$433.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$433.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

W6/32, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

The antibody W6/32 recognises MHC Class I molecules (MHC Class Ia) that are expressed on the surface of all human nucleated cell types. The antibody W6/32 is a valuable reagent for analysing variations in HLA class I expression in different disease states e.g. liver disease, muscular dystrophy, inflammatory myopathy and other neuromuscular disorders.

The previously assigned protein identifier P01891 has been merged into P04439. Full details can be found on the UniProt database.

Immunogen

Membrane of human tonsil cells

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1:300. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Hossein Ashrafi et al.
International journal of cancer, 113(2), 276-283 (2004-09-24)
Papillomaviruses have evolved mechanisms that result in escape from host immune surveillance. The E5 protein is expressed early in papillomavirus infection in the deep layers of the infected epithelium. It is localized to the Golgi apparatus (GA) and endoplasmic reticulum.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service